Assay Details
| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213929.2 | NP_999094.1 | 1 - 2 | 189 | 64 | |
| GenBank mRNA | AF001169.1 | - | 1 - 2 | 118 | 64 | |
| AK232618.1 | - | 1 - 2 | 189 | 64 | ||
| AK350477.1 | - | 1 - 2 | 189 | 64 | ||
| AK397343.1 | - | 1 - 2 | 177 | 64 | ||
| AY609483.1 | - | 1 - 2 | 161 | 64 | ||
| EU093092.1 | - | 1 - 2 | 99 | 64 | ||
| EU571212.1 | - | 1 - 2 | 189 | 64 | ||
| EU571213.1 | - | 1 - 2 | 96 | 64 | ||
| EU571214.1 | - | 1 - 2 | 96 | 64 | ||
| FS687445.1 | - | 1 - 2 | 129 | 64 | ||
More Information
Set Membership: |
|

CTSH
Genomic Map