Assay Details
| Gene Symbol: RPS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213940.1 | NP_999105.1 | 2 - 3 | 134 | 75 | |
| GenBank mRNA | AB000911.1 | - | 2 - 3 | 134 | 75 | |
| AK344436.1 | - | 2 - 3 | 148 | 75 | ||
| AY610361.1 | - | 2 - 3 | 140 | 75 | ||
More Information
Set Membership: |
|

RPS18
Genomic Map