Assay Details
| Gene Symbol: DAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213944.1 | NP_999109.1 | 1 - 2 | 264 | 77 | |
| GenBank mRNA | AK349909.1 | - | 1 - 2 | 272 | 77 | |
| AK391622.1 | - | 1 - 2 | 274 | 77 | ||
| AK394580.1 | - | 1 - 2 | 272 | 77 | ||
| AY609509.1 | - | 1 - 2 | 312 | 77 | ||
| D86562.1 | - | 1 - 2 | 264 | 77 | ||
More Information
Set Membership: |
|

DAD1
Genomic Map