Assay Details
Gene Symbol: PLAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213945.1 | NP_999110.1 | 8 - 9 | 971 | 74 | |
GenBank mRNA | AK391242.1 | - | 8 - 9 | 987 | 74 | |
X02724.1 | - | 971 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
