Assay Details
| Gene Symbol: SLC35A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213951.1 | NP_999116.1 | 1 | 849 | 82 | |
| XM_021080759.1 | XP_020936418.1 | 1429 | 82 | |||
| GenBank mRNA | AJ489473.1 | - | 1 | 849 | 82 | |
| AK398226.1 | - | 1 | 198 | 82 | ||
More Information
Set Membership: |
|

SLC35A4
Genomic Map