Assay Details
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213954.1 | NP_999119.1 | 12 - 13 | 1510 | 98 | |
| GenBank mRNA | AK234325.1 | - | 8 - 9 | 825 | 98 | |
| AK392093.1 | - | 12 - 13 | 1511 | 98 | ||
| J05224.1 | - | 12 - 13 | 1510 | 98 | ||
More Information
Set Membership: |
|

ACO2
Genomic Map