Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213959.1 | NP_999124.1 | 3 - 4 | 926 | 150 | |
| XM_021088684.1 | XP_020944343.1 | 1346 | 150 | |||
| XM_021088685.1 | XP_020944344.1 | 1345 | 150 | |||
| XM_021088686.1 | XP_020944345.1 | 1973 | 150 | |||
| XM_021088687.1 | XP_020944346.1 | 419 | 150 | |||
| XM_021088688.1 | XP_020944347.1 | 628 | 150 | |||
| GenBank mRNA | AF233227.1 | - | 3 - 4 | 926 | 150 | |
More Information
Set Membership: |
|

ANGPT1
Genomic Map