Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213997.1 | NP_999162.1 | 3 - 4 | 217 | 107 | |
| XM_005667326.2 | XP_005667383.1 | 443 | 107 | |||
| XM_005667327.2 | XP_005667384.1 | 414 | 107 | |||
| GenBank mRNA | AB010003.1 | - | 3 - 4 | 217 | 107 | |
| AF176949.1 | - | 3 - 4 | 217 | 107 | ||
| AF191088.1 | - | 3 - 4 | 217 | 107 | ||
| AK391780.1 | - | 4 - 5 | 382 | 107 | ||
| AY450287.1 | - | 3 - 4 | 223 | 107 | ||
| DQ499825.1 | - | 3 - 4 | 217 | 107 | ||
| EF522116.1 | - | 3 - 4 | 217 | 107 | ||
| EU118362.1 | - | 3 - 4 | 217 | 107 | ||
| JF906516.1 | - | 3 - 4 | 217 | 107 | ||
| U68701.1 | - | 3 - 4 | 217 | 107 | ||
| Y11132.2 | - | 217 | 107 | |||
More Information
Set Membership: |
|

IL18
Genomic Map