Assay Details
| Gene Symbol: PRKAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214026.1 | NP_999191.1 | 7 - 8 | 790 | 68 | |
| XM_005656932.3 | XP_005656989.1 | 1161 | 68 | |||
| XM_021066213.1 | XP_020921872.1 | 718 | 68 | |||
| GenBank mRNA | AK238988.1 | - | 7 - 8 | 875 | 68 | |
| AK351482.1 | - | 7 - 8 | 815 | 68 | ||
| AK351943.1 | - | 7 - 8 | 838 | 68 | ||
| AK395800.1 | - | 7 - 8 | 855 | 68 | ||
| AK399335.1 | - | 7 - 8 | 853 | 68 | ||
| X05942.1 | - | 790 | 68 | |||
More Information
Set Membership: |
|

PRKAR1A
Genomic Map