Assay Details
| Gene Symbol: GSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214050.2 | NP_999215.1 | 3 - 4 | 443 | 92 | |
| GenBank mRNA | AF188838.3 | - | 3 - 4 | 446 | 92 | |
| AK235250.1 | - | 3 - 4 | 595 | 92 | ||
| AK391766.1 | - | 3 - 4 | 470 | 92 | ||
| AY609655.1 | - | 3 - 4 | 592 | 92 | ||
More Information
Set Membership: |
|

GSTO1
Genomic Map