Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214374.2 | NP_999539.2 | 4 - 5 | 432 | 68 | |
| GenBank mRNA | AK347117.1 | - | 4 - 5 | 592 | 68 | |
| AK347215.1 | - | 4 - 5 | 758 | 68 | ||
| AK347231.1 | - | 4 - 5 | 442 | 68 | ||
| AK394612.1 | - | 5 - 6 | 447 | 68 | ||
| AK394636.1 | - | 4 - 5 | 388 | 68 | ||
| AK394638.1 | - | 5 - 6 | 416 | 68 | ||
| AK394642.1 | - | 4 - 5 | 601 | 68 | ||
| AK394655.1 | - | 4 - 5 | 419 | 68 | ||
| AK394666.1 | - | 4 - 5 | 419 | 68 | ||
| AK394667.1 | - | 4 - 5 | 422 | 68 | ||
| AK394722.1 | - | 4 - 5 | 423 | 68 | ||
| AK399622.1 | - | 4 - 5 | 421 | 68 | ||
| AK399637.1 | - | 4 - 5 | 419 | 68 | ||
| AY610387.1 | - | 4 - 5 | 555 | 68 | ||
| AY680665.1 | - | 4 - 5 | 440 | 68 | ||
| AY680667.1 | - | 4 - 5 | 440 | 68 | ||
| AY680668.1 | - | 4 - 5 | 440 | 68 | ||
| AY680669.1 | - | 4 - 5 | 440 | 68 | ||
| AY864611.1 | - | 4 - 5 | 432 | 68 | ||
| AY864615.1 | - | 5 - 6 | 462 | 68 | ||
| AY864616.1 | - | 4 - 5 | 401 | 68 | ||
| AY894693.1 | - | 4 - 5 | 432 | 68 | ||
| DQ629158.1 | - | 4 - 5 | 599 | 68 | ||
| DQ629159.1 | - | 4 - 5 | 421 | 68 | ||
| DQ629160.1 | - | 5 - 6 | 448 | 68 | ||
| GU195782.1 | - | 5 - 6 | 449 | 68 | ||
| GU195783.1 | - | 4 - 5 | 390 | 68 | ||
| GU195784.1 | - | 4 - 5 | 385 | 68 | ||
More Information
Set Membership: |
|

MYL1
Genomic Map