Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001626.2 | NP_001001626.2 | 1 - 2 | 257 | 74 | |
| GenBank mRNA | AY533207.1 | - | 1 - 2 | 257 | 74 | |
More Information
Set Membership: |
|

DIO2
Genomic Map