Assay Details
| Gene Symbol: ICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001631.1 | NP_001001631.1 | 2 - 3 | 348 | 65 | |
| GenBank mRNA | AK231951.1 | - | 3 - 4 | 455 | 65 | |
| AK344909.1 | - | 3 - 4 | 670 | 65 | ||
| AK393953.1 | - | 3 - 4 | 462 | 65 | ||
| AK399476.1 | - | 3 - 4 | 483 | 65 | ||
| AY321067.1 | - | 2 - 3 | 348 | 65 | ||
| KU358547.1 | - | 2 - 3 | 326 | 65 | ||
More Information
Set Membership: |
|

ICAM2
Genomic Map