Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001634.1 | NP_001001634.1 | 3 - 4 | 328 | 97 | |
| GenBank mRNA | AK239565.1 | - | 1 | 352 | 97 | |
| AK352159.1 | - | 1327 | 97 | |||
| AY550073.1 | - | 3 - 4 | 328 | 97 | ||
More Information
Set Membership: |
|

RPS17
Genomic Map