Assay Details
| Gene Symbol: SPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001865.1 | NP_001001865.1 | 4 - 5 | 629 | 154 | |
| XM_005660921.3 | XP_005660978.1 | 713 | 154 | |||
| GenBank mRNA | AK232151.1 | - | 4 - 5 | 664 | 154 | |
| AK237187.1 | - | 4 - 5 | 686 | 154 | ||
| AK238919.1 | - | 4 - 5 | 682 | 154 | ||
| AK392995.1 | - | 4 - 5 | 731 | 154 | ||
| AK397628.1 | - | 4 - 5 | 553 | 154 | ||
| AY600299.1 | - | 4 - 5 | 629 | 154 | ||
More Information
Set Membership: |
|

SPI1
Genomic Map