Assay Details
| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004042.1 | NP_001004042.1 | 6 - 7 | 756 | 66 | |
| GenBank mRNA | X62338.1 | - | 756 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1A| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004042.1 | NP_001004042.1 | 6 - 7 | 756 | 66 | |
| GenBank mRNA | X62338.1 | - | 756 | 66 | ||
Set Membership: |
|