Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004046.2 | NP_001004046.1 | 3 - 4 | 330 | 118 | |
| GenBank mRNA | AY960623.1 | - | 3 - 4 | 368 | 118 | |
| EU477533.1 | - | 3 - 4 | 330 | 118 | ||
| X77640.1 | - | 335 | 118 | |||
More Information
Set Membership: |
|

FABP1
Genomic Map