Assay Details
| Gene Symbol: HSD3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004049.2 | NP_001004049.1 | 2 - 3 | 289 | 88 | |
| GenBank mRNA | AB529538.1 | - | 1 - 2 | 147 | 88 | |
| AF232699.2 | - | 2 - 3 | 319 | 88 | ||
| AK235739.1 | - | 2 - 3 | 289 | 88 | ||
| AK235886.1 | - | 2 - 3 | 302 | 88 | ||
| AK236020.1 | - | 1 - 2 | 354 | 88 | ||
| AK348635.1 | - | 2 - 3 | 289 | 88 | ||
More Information
Set Membership: |
|

HSD3B1
Genomic Map