Assay Details
| Gene Symbol: DDX39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005157.1 | NP_001005157.1 | 6 - 7 | 864 | 66 | |
| XM_005665792.3 | XP_005665849.1 | 1100 | 66 | |||
| XM_005665793.3 | XP_005665850.1 | 1087 | 66 | |||
| XM_005665794.3 | XP_005665851.1 | 1151 | 66 | |||
| XM_021098341.1 | XP_020954000.1 | 1531 | 66 | |||
| GenBank mRNA | AK236289.1 | - | 7 - 8 | 1038 | 66 | |
| AK238677.1 | - | 7 - 8 | 1053 | 66 | ||
| AK389641.1 | - | 7 - 8 | 1148 | 66 | ||
| AK389706.1 | - | 7 - 8 | 1055 | 66 | ||
| AK399598.1 | - | 7 - 8 | 1055 | 66 | ||
| EU282340.1 | - | 6 - 7 | 1515 | 66 | ||
More Information
Set Membership: |
|

DDX39B
Genomic Map