Assay Details
| Gene Symbol: ANXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005726.1 | NP_001005726.1 | 9 - 10 | 732 | 60 | |
| XM_005659537.2 | XP_005659594.1 | 918 | 60 | |||
| XM_013992912.2 | XP_013848366.1 | 937 | 60 | |||
| GenBank mRNA | AK231767.1 | - | 9 - 10 | 740 | 60 | |
| AK231957.1 | - | 9 - 10 | 921 | 60 | ||
| AK237951.1 | - | 9 - 10 | 734 | 60 | ||
| AK389658.1 | - | 9 - 10 | 732 | 60 | ||
| AK393851.1 | - | 9 - 10 | 739 | 60 | ||
| AK395129.1 | - | 9 - 10 | 733 | 60 | ||
| AK398743.1 | - | 9 - 10 | 733 | 60 | ||
| AK398851.1 | - | 9 - 10 | 735 | 60 | ||
| AK399603.1 | - | 10 - 11 | 941 | 60 | ||
| AY706383.1 | - | 9 - 10 | 732 | 60 | ||
More Information
Set Membership: |
|

ANXA2
Genomic Map