Assay Details
| Gene Symbol: IL17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005729.1 | NP_001005729.1 | 2 - 3 | 279 | 97 | |
| GenBank mRNA | AB102693.1 | - | 2 - 3 | 279 | 97 | |
| FJ196861.1 | - | 2 - 3 | 247 | 97 | ||
| KF646141.1 | - | 2 - 3 | 233 | 97 | ||
More Information
Set Membership: |
|

IL17A
Genomic Map