Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007192.1 | NP_001007193.1 | 4 - 5 | 643 | 132 | |
| XM_005664110.3 | XP_005664167.1 | 907 | 132 | |||
| XM_021091197.1 | XP_020946856.1 | 987 | 132 | |||
| GenBank mRNA | AY452711.1 | - | 1 - 2 | 162 | 132 | |
| AY606803.1 | - | 4 - 5 | 643 | 132 | ||
More Information
Set Membership: |
|

ADIPOR2
Genomic Map