Assay Details
| Gene Symbol: PRNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008687.1 | NP_001008687.1 | 1 | 652 | 81 | |
| XM_005672669.2 | XP_005672726.1 | 941 | 81 | |||
| XM_013985024.2 | XP_013840478.1 | 889 | 81 | |||
| XM_013985025.2 | XP_013840479.1 | 776 | 81 | |||
| GenBank mRNA | AK235358.1 | - | 2 | 712 | 81 | |
More Information
Set Membership: |
|

PRNP
Genomic Map