Assay Details
| Gene Symbol: MSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009578.1 | NP_001009578.1 | 10 - 11 | 1359 | 57 | |
| XM_013986219.2 | XP_013841673.1 | 1431 | 57 | |||
| GenBank mRNA | M86450.1 | - | 10 - 11 | 1359 | 57 | |
More Information
Set Membership: |
|

MSN
Genomic Map