Assay Details
| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024589.1 | NP_001019760.1 | 3 - 4 | 304 | 100 | |
| XM_005672261.3 | XP_005672318.1 | 339 | 100 | |||
| GenBank mRNA | AJ577084.1 | - | 3 - 4 | 304 | 100 | |
More Information
Set Membership: |
|

CCL20
Genomic Map