Assay Details
| Gene Symbol: MYOZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025222.2 | NP_001020393.1 | 5 - 6 | 900 | 80 | |
| XM_021072179.1 | XP_020927838.1 | 695 | 80 | |||
| XM_021072180.1 | XP_020927839.1 | 2020 | 80 | |||
| GenBank mRNA | DQ058294.1 | - | 4 - 5 | 677 | 80 | |
| DQ143040.1 | - | 5 - 6 | 900 | 80 | ||
More Information
Set Membership: |
|

MYOZ1
Genomic Map