Assay Details
| Gene Symbol: LOC595122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031792.2 | NP_001026962.1 | 1 | 49 | 67 | |
| GenBank mRNA | AK392878.1 | - | 1 | 104 | 67 | |
| AY489288.1 | - | 1 | 49 | 67 | ||
| AY489289.1 | - | 1 | 49 | 67 | ||
More Information
Set Membership: |
|

LOC595122
Genomic Map