Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031794.1 | NP_001026964.1 | 8 - 9 | 876 | 101 | |
| GenBank mRNA | AK392069.1 | - | 9 - 10 | 958 | 101 | |
| AK392352.1 | - | 9 - 10 | 938 | 101 | ||
| AK399497.1 | - | 6 - 7 | 712 | 101 | ||
| AK400063.1 | - | 9 - 10 | 986 | 101 | ||
| AY963262.1 | - | 8 - 9 | 876 | 101 | ||
More Information
Set Membership: |
|

SPARC
Genomic Map