Assay Details
| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033014.1 | NP_001028186.1 | 2 - 3 | 420 | 64 | |
| XM_005653726.3 | XP_005653783.1 | 2469 | 64 | |||
| GenBank mRNA | AK351654.1 | - | 2 - 3 | 419 | 64 | |
| AK394693.1 | - | 2 - 3 | 420 | 64 | ||
| DQ003208.1 | - | 2 - 3 | 420 | 64 | ||
More Information
Set Membership: |
|

MYBPH
Genomic Map