Assay Details
| Gene Symbol: PSMB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037961.1 | NP_001033050.1 | 2 - 3 | 317 | 147 | |
| GenBank mRNA | AK231242.1 | - | 2 - 3 | 163 | 147 | |
| AK392820.1 | - | 2 - 3 | 386 | 147 | ||
| DQ295031.1 | - | 2 - 3 | 317 | 147 | ||
| DQ333333.1 | - | 2 - 3 | 175 | 147 | ||
More Information
Set Membership: |
|

PSMB9
Genomic Map