Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037966.1 | NP_001033055.1 | 1 - 2 | 202 | 94 | |
| GenBank mRNA | DQ132880.1 | - | 1 - 2 | 202 | 94 | |
More Information
Set Membership: |
|

TEAD3
Genomic Map