Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038631.1 | NP_001033720.1 | 2 - 3 | 306 | 99 | |
| GenBank mRNA | AK231712.1 | - | 2 - 3 | 274 | 99 | |
| AK231896.1 | - | 2 - 3 | 288 | 99 | ||
| AK348546.1 | - | 2 - 3 | 254 | 99 | ||
| AK352130.1 | - | 2 - 3 | 227 | 99 | ||
| AK392435.1 | - | 2 - 3 | 289 | 99 | ||
| AK399226.1 | - | 2 - 3 | 286 | 99 | ||
| AY857634.1 | - | 2 - 3 | 306 | 99 | ||
More Information
Set Membership: |
|

PTGES
Genomic Map