Assay Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039746.2 | NP_001034835.1 | 1 - 2 | 159 | 86 | |
| GenBank mRNA | AB529462.1 | - | 1 - 2 | 77 | 86 | |
| AK231381.1 | - | 1 - 2 | 160 | 86 | ||
| AK346851.1 | - | 317 | 86 | |||
| AY841270.1 | - | 1 - 2 | 162 | 86 | ||
| DQ372083.1 | - | 1 - 2 | 160 | 86 | ||
| DQ523617.1 | - | 1 - 2 | 44 | 86 | ||
More Information
Set Membership: |
|

FABP5
Genomic Map