Assay Details
| Gene Symbol: MTX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039750.1 | NP_001034839.1 | 3 - 4 | 239 | 69 | |
| GenBank mRNA | AB239841.1 | - | 3 - 4 | 250 | 69 | |
| AK234275.1 | - | 3 - 4 | 250 | 69 | ||
| AK347776.1 | - | 3 - 4 | 226 | 69 | ||
| AK392795.1 | - | 3 - 4 | 243 | 69 | ||
| DQ402035.1 | - | 3 - 4 | 239 | 69 | ||
More Information
Set Membership: |
|

MTX1
Genomic Map