Assay Details
| Gene Symbol: TRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375395.1 | NP_001362324.1 | 12 | 6345 | 84 | |
| NM_001375396.1 | NP_001362325.1 | 11 | 5236 | 84 | ||
| NM_213741.3 | NP_998906.2 | 12 | 6391 | 84 | ||
| XM_021079491.1 | XP_020935150.1 | 5329 | 84 | |||
| GenBank mRNA | AB081619.1 | - | 1 | 1549 | 84 | |
More Information
Set Membership: |
|

TRO
Genomic Map