Assay Details
| Gene Symbol: MSMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213752.1 | NP_998917.1 | 2 - 3 | 421 | 116 | |
| XM_005656503.3 | XP_005656560.1 | 116 | ||||
| GenBank mRNA | AB530151.1 | - | 2 - 3 | 116 | ||
| AK235870.1 | - | 3 - 4 | 116 | |||
| AK391239.1 | - | 3 - 4 | 116 | |||
| AY370690.1 | - | 2 - 3 | 421 | 116 | ||
More Information
Set Membership: |
|

MSMO1
Genomic Map