Assay Details
| Gene Symbol: TSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213753.1 | NP_998918.1 | 3 - 4 | 387 | 100 | |
| GenBank mRNA | AY366497.1 | - | 3 - 4 | 387 | 100 | |
| AY466517.1 | - | 2 - 3 | 176 | 100 | ||
| AY644722.1 | - | 3 - 4 | 303 | 100 | ||
More Information
Set Membership: |
|

TSPO
Genomic Map