Assay Details
| Gene Symbol: DSPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100402.1 | XP_020956061.1 | 1721 | 115 | ||
| XM_021100403.1 | XP_020956062.1 | 1636 | 115 | |||
| XM_021100404.1 | XP_020956063.1 | 1494 | 115 | |||
| XM_021100405.1 | XP_020956064.1 | 1403 | 115 | |||
| XM_021100406.1 | XP_020956065.1 | 1716 | 115 | |||
| GenBank mRNA | AY161862.1 | - | 4 - 5 | 1245 | 115 | |
| AY161863.1 | - | 4 - 5 | 1255 | 115 | ||
More Information
Set Membership: |
|

DSPP
Genomic Map