Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213781.1 | NP_998946.1 | 2 - 3 | 483 | 65 | |
| XM_021072070.1 | XP_020927729.1 | 731 | 65 | |||
| GenBank mRNA | AK344093.1 | - | 2 - 3 | 483 | 65 | |
| AK344131.1 | - | 2 - 3 | 483 | 65 | ||
| AK399498.1 | - | 2 - 3 | 483 | 65 | ||
| AY487829.1 | - | 2 - 3 | 483 | 65 | ||
| JN613287.1 | - | 2 - 3 | 380 | 65 | ||
| Z97186.1 | - | 275 | 65 | |||
More Information
Set Membership: |
|

SCD
Genomic Map