Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213808.1 | NP_998973.1 | 5 - 6 | 1253 | 69 | |
| XM_005671650.3 | XP_005671707.1 | 7956 | 69 | |||
| GenBank mRNA | AF233228.1 | - | 5 - 6 | 1253 | 69 | |
| AY616677.1 | - | 2 - 3 | 195 | 69 | ||
More Information
Set Membership: |
|

ANGPT2
Genomic Map