Assay Details
| Gene Symbol: PPAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213809.1 | NP_998974.1 | 6 - 7 | 771 | 115 | |
| GenBank mRNA | AF315377.1 | - | 6 - 7 | 771 | 115 | |
| AY188554.1 | - | 5 - 6 | 630 | 115 | ||
More Information
Set Membership: |
|

PPAG3
Genomic Map