Assay Details
| Gene Symbol: SAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213814.2 | NP_998979.1 | 6 - 7 | 622 | 78 | |
| GenBank mRNA | AJ249974.1 | - | 6 - 7 | 622 | 78 | |
| AK350053.1 | - | 6 - 7 | 656 | 78 | ||
| AK399206.1 | - | 6 - 7 | 648 | 78 | ||
More Information
Set Membership: |
|

SAL1
Genomic Map