Assay Details
| Gene Symbol: CCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213833.2 | NP_998998.2 | 2 - 3 | 510 | 72 | |
| GenBank mRNA | AK346093.1 | - | 2 - 3 | 511 | 72 | |
| U70060.1 | - | 2 - 3 | 72 | |||
| U83916.1 | - | 2 - 3 | 499 | 72 | ||
More Information
Set Membership: |
|

CCN2
Genomic Map