Assay Details
| Gene Symbol: AMCF-II | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213876.1 | NP_999041.1 | 1 - 2 | 148 | 92 | |
| GenBank mRNA | AK344686.1 | - | 1 - 2 | 211 | 92 | |
| M99368.1 | - | 1 - 2 | 148 | 92 | ||
More Information
Set Membership: |
|

AMCF-II
Genomic Map