Assay Details
| Gene Symbol: CNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213878.1 | NP_999043.1 | 4 - 5 | 463 | 83 | |
| XM_013987363.2 | XP_013842817.1 | 412 | 83 | |||
| XM_021080565.1 | XP_020936224.1 | 440 | 83 | |||
| GenBank mRNA | Z19538.1 | - | 463 | 83 | ||
More Information
Set Membership: |
|

CNN1
Genomic Map