Assay Details
| Gene Symbol: PROC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213918.1 | NP_999083.1 | 8 | 1357 | 62 | |
| XM_005671629.2 | XP_005671686.1 | 1485 | 62 | |||
| XM_005671630.3 | XP_005671687.1 | 1509 | 62 | |||
| XM_013983970.2 | XP_013839424.1 | 1608 | 62 | |||
| XM_013983971.2 | XP_013839425.1 | 1402 | 62 | |||
| XM_021074671.1 | XP_020930330.1 | 1582 | 62 | |||
| XM_021074672.1 | XP_020930331.1 | 1569 | 62 | |||
| XM_021074673.1 | XP_020930332.1 | 1321 | 62 | |||
| GenBank mRNA | AF191307.1 | - | 8 | 1357 | 62 | |
More Information
Set Membership: |
|

PROC
Genomic Map