Assay Details
| Gene Symbol: ALOX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213931.1 | NP_999096.1 | 13 | 1749 | 72 | |
| XM_013981286.2 | XP_013836740.2 | 2530 | 72 | |||
| GenBank mRNA | AK390768.1 | - | 1 | 1410 | 72 | |
| M31417.1 | - | 13 | 1749 | 72 | ||
More Information
Set Membership: |
|

ALOX15
Genomic Map