Assay Details
| Gene Symbol: ARSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213933.1 | NP_999098.1 | 9 | 1652 | 58 | |
| GenBank mRNA | AF316108.2 | - | 9 | 1652 | 58 | |
| AK232471.1 | - | 9 | 1496 | 58 | ||
| AK234774.1 | - | 9 | 1543 | 58 | ||
| AK389312.1 | - | 9 | 1566 | 58 | ||
More Information
Set Membership: |
|

ARSA
Genomic Map