Assay Details
| Gene Symbol: PDXK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213943.1 | NP_999108.1 | 8 - 9 | 647 | 144 | |
| GenBank mRNA | AF041255.1 | - | 8 - 9 | 676 | 144 | |
| AK234117.1 | - | 8 - 9 | 775 | 144 | ||
| AK235762.1 | - | 8 - 9 | 775 | 144 | ||
| AK352196.1 | - | 8 - 9 | 775 | 144 | ||
More Information
Set Membership: |
|

PDXK
Genomic Map