Assay Details
| Gene Symbol: RPL29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213950.1 | NP_999115.1 | 4 | 448 | 113 | |
| XM_005669562.3 | XP_005669619.1 | 483 | 113 | |||
| GenBank mRNA | AB016798.1 | - | 4 | 448 | 113 | |
| AK236091.1 | - | 4 | 455 | 113 | ||
| AK392098.1 | - | 4 | 451 | 113 | ||
More Information
Set Membership: |
|

RPL29
Genomic Map